Assay Details
| Gene Symbol: CUNH6orf136 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004847086.3 | XP_004847143.1 | 4 - 5 | 935 | 80 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

CUNH6orf136| Gene Symbol: CUNH6orf136 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004847086.3 | XP_004847143.1 | 4 - 5 | 935 | 80 | |