Assay Details
| Gene Symbol: Cry2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004851964.3 | XP_004852021.1 | 11 - 12 | 1824 | 69 | |
| XM_021264296.1 | XP_021119955.1 | 12 - 13 | 1842 | 69 | ||
More Information
Set Membership: |
|

Cry2
Genomic Map