Assay Details
| Gene Symbol: Alox5ap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004854781.2 | XP_004854838.1 | 2 - 3 | 238 | 61 | |
| XM_021236309.1 | XP_021091968.1 | 4 - 5 | 584 | 61 | ||
| XM_021236310.1 | XP_021091969.1 | 3 - 4 | 418 | 61 | ||
| XM_021236311.1 | XP_021091970.1 | 3 - 4 | 360 | 61 | ||
More Information
Set Membership: |
|

Alox5ap
Genomic Map