Assay Details
| Gene Symbol: Islr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004855478.3 | XP_004855535.1 | 3 - 4 | 470 | 83 | |
| XM_004855479.3 | XP_004855536.1 | 2 - 3 | 363 | 83 | ||
| XM_004855480.3 | XP_004855537.1 | 2 - 3 | 239 | 83 | ||
| XM_021237116.1 | XP_021092775.1 | 2 - 3 | 1019 | 83 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_004855478.3, XM_004855479.3, XM_004855480.3 transcripts.
Set Membership: |
|

Islr2
Genomic Map