Assay Details
| Gene Symbol: LOC101703542 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004860824.3 | XP_004860881.1 | 6 - 7 | 1252 | 82 | |
| XM_021241142.1 | XP_021096801.1 | 6 - 7 | 1438 | 82 | ||
| XM_021241143.1 | XP_021096802.1 | 6 - 7 | 1300 | 82 | ||
| XM_021241144.1 | XP_021096803.1 | 5 - 6 | 1040 | 82 | ||
More Information
Set Membership: |
|

LOC101703542
Genomic Map