Assay Details
| Gene Symbol: Pgs1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004860828.2 | XP_004860885.1 | 6 - 7 | 909 | 59 | |
| XM_021241336.1 | XP_021096995.1 | 8 - 9 | 1074 | 59 | ||
| XM_021241337.1 | XP_021096996.1 | 6 - 7 | 909 | 59 | ||
| XM_021241338.1 | XP_021096997.1 | 6 - 7 | 909 | 59 | ||
| XM_021241339.1 | XP_021096998.1 | 7 - 8 | 977 | 59 | ||
More Information
Set Membership: |
|

Pgs1
Genomic Map