Assay Details
| Gene Symbol: Pip4k2c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004861199.3 | XP_004861256.1 | 9 - 10 | 1268 | 67 | |
| XM_013078295.2 | XP_012933749.1 | 8 - 9 | 1230 | 67 | ||
| XM_021241408.1 | XP_021097067.1 | 9 - 10 | 1027 | 67 | ||
More Information
Set Membership: |
|

Pip4k2c
Genomic Map