Assay Details
| Gene Symbol: Cd180 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004862843.3 | XP_004862900.1 | 1 - 2 | 233 | 107 | |
| XM_021242977.1 | XP_021098636.1 | 1 - 2 | 233 | 107 | ||
| XM_021242978.1 | XP_021098637.1 | 1 - 2 | 434 | 107 | ||
More Information
Set Membership: |
|

Cd180
Genomic Map