Assay Details
| Gene Symbol: LOC101705747 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004864215.3 | XP_004864272.3 | 4 - 5 | 482 | 56 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101705747| Gene Symbol: LOC101705747 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004864215.3 | XP_004864272.3 | 4 - 5 | 482 | 56 | |
Set Membership: |
|