Assay Details
| Gene Symbol: LOC101704584 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013078863.1 | XP_012934317.1 | 3 - 4 | 425 | 89 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101704584| Gene Symbol: LOC101704584 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013078863.1 | XP_012934317.1 | 3 - 4 | 425 | 89 | |
Set Membership: |
|