Assay Details
| Gene Symbol: CUNH12orf40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021236513.1 | XP_021092172.1 | 10 - 11 | 1694 | 100 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

CUNH12orf40| Gene Symbol: CUNH12orf40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021236513.1 | XP_021092172.1 | 10 - 11 | 1694 | 100 | |