Assay Details
| Gene Symbol: CUNH12orf40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021236513.1 | XP_021092172.1 | 7 - 8 | 886 | 65 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CUNH12orf40| Gene Symbol: CUNH12orf40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021236513.1 | XP_021092172.1 | 7 - 8 | 886 | 65 | |
Set Membership: |
|