Assay Details
| Gene Symbol: LOC101723046 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021247995.1 | XP_021103654.1 | 13 - 14 | 1774 | 101 | |
| XM_021248003.1 | XP_021103662.1 | 13 - 14 | 1772 | 101 | ||
| XM_021248008.1 | XP_021103667.1 | 8 - 9 | 1111 | 101 | ||
| XM_021248010.1 | XP_021103669.1 | 13 - 14 | 1777 | 101 | ||
| XM_021248024.1 | XP_021103683.1 | 13 - 14 | 1779 | 101 | ||
More Information
Set Membership: |
|

LOC101723046
Genomic Map