Assay Details
| Gene Symbol: LOC101723046 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021247995.1 | XP_021103654.1 | 16 - 17 | 2146 | 135 | |
| XM_021248003.1 | XP_021103662.1 | 16 - 17 | 2144 | 135 | ||
| XM_021248008.1 | XP_021103667.1 | 11 - 12 | 1483 | 135 | ||
| XM_021248010.1 | XP_021103669.1 | 16 - 17 | 2149 | 135 | ||
| XM_021248024.1 | XP_021103683.1 | 16 - 17 | 2151 | 135 | ||
More Information
Set Membership: |
|

LOC101723046
Genomic Map