Assay Details
| Gene Symbol: LOC101723046 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021247995.1 | XP_021103654.1 | 20 - 21 | 2796 | 97 | |
| XM_021248003.1 | XP_021103662.1 | 20 - 21 | 2794 | 97 | ||
| XM_021248008.1 | XP_021103667.1 | 15 - 16 | 2133 | 97 | ||
| XM_021248010.1 | XP_021103669.1 | 20 - 21 | 2799 | 97 | ||
| XM_021248024.1 | XP_021103683.1 | 20 - 21 | 2801 | 97 | ||

LOC101723046
Genomic Map