Assay Details
| Gene Symbol: LOC101723046 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021247995.1 | XP_021103654.1 | 29 - 30 | 4195 | 131 | |
| XM_021248003.1 | XP_021103662.1 | 29 - 30 | 4193 | 131 | ||
| XM_021248008.1 | XP_021103667.1 | 24 - 25 | 3532 | 131 | ||
More Information
Set Membership: |
|

LOC101723046
Genomic Map