Assay Details
| Gene Symbol: LOC101723046 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021247995.1 | XP_021103654.1 | 33 - 34 | 4713 | 93 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101723046| Gene Symbol: LOC101723046 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021247995.1 | XP_021103654.1 | 33 - 34 | 4713 | 93 | |
Set Membership: |
|