Assay Details
| Gene Symbol: LOC101723046 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021247995.1 | XP_021103654.1 | 9 - 10 | 1154 | 88 | |
| XM_021248003.1 | XP_021103662.1 | 9 - 10 | 1152 | 88 | ||
| XM_021248008.1 | XP_021103667.1 | 4 - 5 | 491 | 88 | ||
| XM_021248010.1 | XP_021103669.1 | 9 - 10 | 1157 | 88 | ||
| XM_021248024.1 | XP_021103683.1 | 9 - 10 | 1159 | 88 | ||

LOC101723046
Genomic Map