Assay Details
| Gene Symbol: LOC101703188 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021253061.1 | XP_021108720.1 | 2 - 3 | 443 | 94 | |
| XM_021253062.1 | XP_021108721.1 | 2 - 3 | 441 | 94 | ||
| XM_021253068.1 | XP_021108727.1 | 1 - 2 | 52 | 94 | ||
| XM_021253070.1 | XP_021108729.1 | 2 - 3 | 444 | 94 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021253068.1 transcripts.
Set Membership: |
|

LOC101703188
Genomic Map