Assay Details
| Gene Symbol: Zzz3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021255497.1 | XP_021111156.1 | 13 - 14 | 2934 | 67 | |
| XM_021255498.1 | XP_021111157.1 | 13 - 14 | 3073 | 67 | ||
| XM_021255499.1 | XP_021111158.1 | 14 - 15 | 2993 | 67 | ||
| XM_021255500.1 | XP_021111159.1 | 12 - 13 | 2781 | 67 | ||
| XM_021255501.1 | XP_021111160.1 | 13 - 14 | 2931 | 67 | ||
| XM_021255502.1 | XP_021111161.1 | 12 - 13 | 1378 | 67 | ||
More Information
Set Membership: |
|

Zzz3
Genomic Map