Assay Details
| Gene Symbol: Basp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021258634.1 | XP_021114293.1 | 1 - 2 | 87 | 84 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Basp1| Gene Symbol: Basp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021258634.1 | XP_021114293.1 | 1 - 2 | 87 | 84 | |
Set Membership: |
|