Assay Details
| Gene Symbol: Slc33a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004834497.3 | XP_004834554.1 | 5 - 6 | 2087 | 71 | |
| XM_021253442.1 | XP_021109101.1 | 5 - 6 | 2086 | 71 | ||
| XM_021253450.1 | XP_021109109.1 | 4 - 5 | 657 | 71 | ||
More Information
Set Membership: |
|

Slc33a1
Genomic Map