Assay Details
| Gene Symbol: Mepce | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004840209.3 | XP_004840266.1 | 1 - 2 | 2441 | 73 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mepce| Gene Symbol: Mepce | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004840209.3 | XP_004840266.1 | 1 - 2 | 2441 | 73 | |