Assay Details
| Gene Symbol: Mepce | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004840209.3 | XP_004840266.1 | 2 - 3 | 2660 | 71 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mepce| Gene Symbol: Mepce | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004840209.3 | XP_004840266.1 | 2 - 3 | 2660 | 71 | |
Set Membership: |
|