Assay Details
| Gene Symbol: Erg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004842372.2 | XP_004842429.1 | 11 - 12 | 1148 | 76 | |
| XM_004842373.3 | XP_004842430.1 | 9 - 10 | 1044 | 76 | ||
| XM_021256245.1 | XP_021111904.1 | 9 - 10 | 1238 | 76 | ||
| XM_021256246.1 | XP_021111905.1 | 8 - 9 | 1169 | 76 | ||
| XM_021256247.1 | XP_021111906.1 | 7 - 8 | 1089 | 76 | ||
More Information
Set Membership: |
|

Erg
Genomic Map