Assay Details
| Gene Symbol: Bace2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004842466.2 | XP_004842523.1 | 7 - 8 | 1123 | 61 | |
| XM_021256354.1 | XP_021112013.1 | 7 - 8 | 924 | 61 | ||
More Information
Set Membership: |
|

Bace2
Genomic Map