Assay Details
Gene Symbol: Nol7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004847664.3 | XP_004847721.1 | 2 - 3 | 384 | 62 | |
XM_021260638.1 | XP_021116297.1 | 2 - 3 | 384 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
