Assay Details
| Gene Symbol: Pgbd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004847836.3 | XP_004847893.2 | 2 - 3 | 531 | 85 | |
| XM_013073172.2 | XP_012928626.1 | 4 - 5 | 1094 | 85 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pgbd1| Gene Symbol: Pgbd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004847836.3 | XP_004847893.2 | 2 - 3 | 531 | 85 | |
| XM_013073172.2 | XP_012928626.1 | 4 - 5 | 1094 | 85 | ||