Assay Details
| Gene Symbol: Pgbd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004847836.3 | XP_004847893.2 | 4 - 5 | 715 | 78 | |
| XM_013073172.2 | XP_012928626.1 | 6 - 7 | 1278 | 78 | ||
| XM_021260778.1 | XP_021116437.1 | 7 - 8 | 1334 | 78 | ||
| XM_021260779.1 | XP_021116438.1 | 7 - 8 | 1273 | 78 | ||
| XM_021260780.1 | XP_021116439.1 | 8 - 9 | 1478 | 78 | ||
| XM_021260781.1 | XP_021116440.1 | 7 - 8 | 1364 | 78 | ||
| XM_021260783.1 | XP_021116442.1 | 8 - 9 | 1417 | 78 | ||
| XM_021260784.1 | XP_021116443.1 | 7 - 8 | 1303 | 78 | ||
| XM_021260785.1 | XP_021116444.1 | 6 - 7 | 1258 | 78 | ||
| XM_021260786.1 | XP_021116445.1 | 5 - 6 | 769 | 78 | ||
| XM_021260787.1 | XP_021116446.1 | 5 - 6 | 587 | 78 | ||
| XM_021260788.1 | XP_021116447.1 | 5 - 6 | 735 | 78 | ||
| XM_021260789.1 | XP_021116448.1 | 4 - 5 | 694 | 78 | ||
More Information
Set Membership: |
|

Pgbd1
Genomic Map