Assay Details
| Gene Symbol: Ghr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004848566.3 | XP_004848623.1 | 4 - 5 | 590 | 74 | |
| XM_004848567.3 | XP_004848624.1 | 4 - 5 | 387 | 74 | ||
| XM_004848568.3 | XP_004848625.1 | 4 - 5 | 606 | 74 | ||
| XM_021261444.1 | XP_021117103.1 | 6 - 7 | 840 | 74 | ||
| XM_021261445.1 | XP_021117104.1 | 5 - 6 | 774 | 74 | ||
| XM_021261446.1 | XP_021117105.1 | 5 - 6 | 680 | 74 | ||
| XM_021261447.1 | XP_021117106.1 | 3 - 4 | 525 | 74 | ||
| XM_021261448.1 | XP_021117107.1 | 3 - 4 | 321 | 74 | ||
| XM_021261449.1 | XP_021117108.1 | 3 - 4 | 540 | 74 | ||
| XM_021261450.1 | XP_021117109.1 | 2 - 3 | 161 | 74 | ||
| XM_021261451.1 | XP_021117110.1 | 6 - 7 | 842 | 74 | ||
| XM_021261452.1 | XP_021117111.1 | 6 - 7 | 842 | 74 | ||
More Information
Set Membership: |
|

Ghr
Genomic Map