Assay Details
| Gene Symbol: Hexa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004855522.3 | XP_004855579.1 | 7 - 8 | 842 | 62 | |
| XM_004855523.3 | XP_004855580.1 | 7 - 8 | 842 | 62 | ||
| XM_013076263.2 | XP_012931717.1 | 7 - 8 | 843 | 62 | ||
| XM_021236968.1 | XP_021092627.1 | 7 - 8 | 842 | 62 | ||
| XM_021236969.1 | XP_021092628.1 | 7 - 8 | 741 | 62 | ||
More Information
Set Membership: |
|

Hexa
Genomic Map