Assay Details
| Gene Symbol: Acp7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004859092.2 | XP_004859149.1 | 5 - 6 | 1164 | 62 | |
| XM_021239770.1 | XP_021095429.1 | 5 - 6 | 1167 | 62 | ||
| XM_021239771.1 | XP_021095430.1 | 5 - 6 | 1085 | 62 | ||
More Information
Set Membership: |
|

Acp7
Genomic Map