Assay Details
Gene Symbol: Znf516 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004861935.3 | XP_004861992.1 | 3 - 4 | 3513 | 92 | |
XM_004861936.3 | XP_004861993.1 | 3 - 4 | 3494 | 92 | ||
XM_004861937.3 | XP_004861994.1 | 3 - 4 | 3517 | 92 | ||
XM_004861939.3 | XP_004861996.1 | 3 - 4 | 3513 | 92 | ||
XM_021242177.1 | XP_021097836.1 | 3 - 4 | 4073 | 92 | ||
XM_021242178.1 | XP_021097837.1 | 3 - 4 | 3481 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
