Assay Details
| Gene Symbol: Mmp23b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004863935.2 | XP_004863992.1 | 3 - 4 | 440 | 56 | |
| XM_021243644.1 | XP_021099303.1 | 3 - 4 | 440 | 56 | ||
| XM_021243645.1 | XP_021099304.1 | 3 - 4 | 344 | 56 | ||
| XM_021243646.1 | XP_021099305.1 | 3 - 4 | 440 | 56 | ||
| XM_021243647.1 | XP_021099306.1 | 3 - 4 | 440 | 56 | ||
| XM_021243648.1 | XP_021099307.1 | 3 - 4 | 440 | 56 | ||
More Information
Set Membership: |
|

Mmp23b
Genomic Map