Assay Details
| Gene Symbol: Mmp23b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004863935.2 | XP_004863992.1 | 7 - 8 | 1003 | 100 | |
| XM_021243644.1 | XP_021099303.1 | 8 - 9 | 926 | 100 | ||
| XM_021243645.1 | XP_021099304.1 | 8 - 9 | 830 | 100 | ||
| XM_021243648.1 | XP_021099307.1 | 7 - 8 | 960 | 100 | ||
More Information
Set Membership: |
|

Mmp23b
Genomic Map