Assay Details
| Gene Symbol: Lmx1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004865181.2 | XP_004865238.1 | 8 - 9 | 1164 | 69 | |
| XM_021244720.1 | XP_021100379.1 | 8 - 9 | 1090 | 69 | ||
More Information
Set Membership: |
|

Lmx1a
Genomic Map