Assay Details
| Gene Symbol: Angptl4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004865406.3 | XP_004865463.1 | 3 - 4 | 669 | 72 | |
| XM_004865408.3 | XP_004865465.1 | 3 - 4 | 454 | 72 | ||
More Information
Set Membership: |
|

Angptl4
Genomic Map