Assay Details
| Gene Symbol: Unkl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004873851.3 | XP_004873908.1 | 10 - 11 | 1296 | 64 | |
| XM_013068882.2 | XP_012924336.1 | 8 - 9 | 1125 | 64 | ||
| XM_013068883.2 | XP_012924337.1 | 9 - 10 | 1086 | 64 | ||
| XM_013068884.2 | XP_012924338.1 | 8 - 9 | 882 | 64 | ||
| XM_013068887.2 | XP_012924341.1 | 9 - 10 | 1293 | 64 | ||
| XM_021251369.1 | XP_021107028.1 | 10 - 11 | 1296 | 64 | ||
| XM_021251370.1 | XP_021107029.1 | 8 - 9 | 960 | 64 | ||
| XM_021251371.1 | XP_021107030.1 | 10 - 11 | 1099 | 64 | ||
| XM_021251372.1 | XP_021107031.1 | 6 - 7 | 599 | 64 | ||
More Information
Set Membership: |
|

Unkl
Genomic Map