Assay Details
| Gene Symbol: LOC101713038 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013070279.2 | XP_012925733.1 | 9 - 10 | 888 | 81 | |
| XM_021254096.1 | XP_021109755.1 | 9 - 10 | 858 | 81 | ||
| XM_021254097.1 | XP_021109756.1 | 9 - 10 | 991 | 81 | ||
| XM_021254098.1 | XP_021109757.1 | 9 - 10 | 951 | 81 | ||
More Information
Set Membership: |
|

LOC101713038
Genomic Map