Assay Details
| Gene Symbol: Mybl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013077111.2 | XP_012932565.2 | 8 - 9 | 1574 | 58 | |
| XM_021239070.1 | XP_021094729.1 | 8 - 9 | 1591 | 58 | ||
More Information
Set Membership: |
|

Mybl2
Genomic Map