Assay Details
| Gene Symbol: Cdk3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013078142.2 | XP_012933596.2 | 2 - 3 | 683 | 82 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cdk3| Gene Symbol: Cdk3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013078142.2 | XP_012933596.2 | 2 - 3 | 683 | 82 | |
Set Membership: |
|