Assay Details
| Gene Symbol: Pi4k2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004838609.2 | XP_004838666.1 | 7 - 8 | 1281 | 62 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pi4k2a| Gene Symbol: Pi4k2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004838609.2 | XP_004838666.1 | 7 - 8 | 1281 | 62 | |
Set Membership: |
|