Assay Details
| Gene Symbol: LOC101711381 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004843524.1 | XP_004843581.1 | 12 - 13 | 1452 | 101 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101711381| Gene Symbol: LOC101711381 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004843524.1 | XP_004843581.1 | 12 - 13 | 1452 | 101 | |
Set Membership: |
|