Assay Details
| Gene Symbol: Gpat2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004844349.3 | XP_004844406.1 | 11 - 12 | 1448 | 59 | |
| XM_021258004.1 | XP_021113663.1 | 11 - 12 | 1448 | 59 | ||
| XM_021258005.1 | XP_021113664.1 | 11 - 12 | 1448 | 59 | ||
| XM_021258006.1 | XP_021113665.1 | 11 - 12 | 1448 | 59 | ||
| XM_021258007.1 | XP_021113666.1 | 11 - 12 | 1448 | 59 | ||
| XM_021258008.1 | XP_021113667.1 | 11 - 12 | 1448 | 59 | ||
| XM_021258009.1 | XP_021113668.1 | 11 - 12 | 1448 | 59 | ||
More Information
Set Membership: |
|

Gpat2
Genomic Map