Assay Details
| Gene Symbol: Maff | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004845618.3 | XP_004845675.1 | 2 - 3 | 351 | 61 | |
| XM_013072414.2 | XP_012927868.1 | 2 - 3 | 2214 | 61 | ||
| XM_021259003.1 | XP_021114662.1 | 1 - 2 | 872 | 61 | ||
| XM_021259004.1 | XP_021114663.1 | 2 - 3 | 758 | 61 | ||
| XM_021259005.1 | XP_021114664.1 | 2 - 3 | 3873 | 61 | ||
More Information
Set Membership: |
|

Maff
Genomic Map