Assay Details
| Gene Symbol: Epb41l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004845983.3 | XP_004846040.1 | 7 - 8 | 1310 | 70 | |
| XM_013072659.2 | XP_012928113.1 | 7 - 8 | 1325 | 70 | ||
| XM_013072660.2 | XP_012928114.1 | 7 - 8 | 1324 | 70 | ||
| XM_013072661.2 | XP_012928115.1 | 7 - 8 | 1322 | 70 | ||
| XM_013072662.2 | XP_012928116.1 | 7 - 8 | 1315 | 70 | ||
| XM_013072664.2 | XP_012928118.1 | 7 - 8 | 1305 | 70 | ||
| XM_013072667.2 | XP_012928121.1 | 7 - 8 | 1306 | 70 | ||
| XM_013072668.2 | XP_012928122.1 | 7 - 8 | 1301 | 70 | ||
| XM_021259243.1 | XP_021114902.1 | 7 - 8 | 1215 | 70 | ||
| XM_021259244.1 | XP_021114903.1 | 7 - 8 | 1319 | 70 | ||
| XM_021259245.1 | XP_021114904.1 | 7 - 8 | 1328 | 70 | ||
| XM_021259246.1 | XP_021114905.1 | 7 - 8 | 1309 | 70 | ||
| XM_021259247.1 | XP_021114906.1 | 7 - 8 | 1306 | 70 | ||
| XM_021259248.1 | XP_021114907.1 | 7 - 8 | 1304 | 70 | ||
| XM_021259249.1 | XP_021114908.1 | 7 - 8 | 1309 | 70 | ||
More Information
Set Membership: |
|

Epb41l2
Genomic Map