Assay Details
| Gene Symbol: Ptchd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004847138.3 | XP_004847195.2 | 2 - 3 | 1157 | 66 | |
| XM_021259695.1 | XP_021115354.1 | 2 - 3 | 983 | 66 | ||
More Information
Set Membership: |
|

Ptchd4
Genomic Map