Assay Details
| Gene Symbol: Irf4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004847797.2 | XP_004847854.1 | 4 - 5 | 653 | 70 | |
| XM_004847798.2 | XP_004847855.1 | 4 - 5 | 650 | 70 | ||
More Information
Set Membership: |
|

Irf4
Genomic Map