Assay Details
| Gene Symbol: Prdm11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004851977.3 | XP_004852034.3 | 3 - 4 | 488 | 69 | |
| XM_013074927.2 | XP_012930381.1 | 2 - 3 | 241 | 69 | ||
| XM_013074928.2 | XP_012930382.2 | 3 - 4 | 486 | 69 | ||
| XM_021264302.1 | XP_021119961.1 | 3 - 4 | 460 | 69 | ||
| XM_021264303.1 | XP_021119962.1 | 3 - 4 | 459 | 69 | ||
| XM_021264304.1 | XP_021119963.1 | 3 - 4 | 480 | 69 | ||
| XM_021264305.1 | XP_021119964.1 | 3 - 4 | 456 | 69 | ||
| XM_021264306.1 | XP_021119965.1 | 2 - 3 | 241 | 69 | ||
More Information
Set Membership: |
|

Prdm11
Genomic Map