Assay Details
| Gene Symbol: Gal3st3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004852341.2 | XP_004852398.1 | 2 - 3 | 451 | 118 | |
| XM_004852342.2 | XP_004852399.1 | 2 - 3 | 322 | 118 | ||
| XM_021264559.1 | XP_021120218.1 | 2 - 3 | 521 | 118 | ||
More Information
Set Membership: |
|

Gal3st3
Genomic Map